siRNA Details
| Gene Symbol: Cdc42bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053620.3 | NP_446072.2 | 28 | 3821 | ||
| XM_006240562.2 | XP_006240624.1 | 29 | 4021 | |||
| GenBank mRNA | AF021936.1 | - | 28 | 3781 | ||
| AY277590.1 | - | 28 | 3821 | |||

Cdc42bpb
Genomic Map