siRNA Details
| Gene Symbol: Gorasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007720.1 | NP_001007721.1 | 5 | 511 | ||
| GenBank mRNA | AF110267.1 | - | 5 | 519 | ||
| BC078731.1 | - | 5 | 511 | 7112220 | ||
| Gene Symbol: LOC103690018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761953.1 | XP_008760175.1 | 4 | 493 | ||

Gorasp2
Genomic Map