siRNA Details
| Gene Symbol: Gorasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007720.1 | NP_001007721.1 | 7, 8 | 842 | ||
| GenBank mRNA | BC078731.1 | - | 7, 8 | 842 | 7112220 | |
| Gene Symbol: LOC103690018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761953.1 | XP_008760175.1 | 6, 7 | 824 | ||

Gorasp2
Genomic Map