siRNA Details
| Gene Symbol: Pitx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053624.2 | NP_446076.1 | 3 | 1165 | ||
| XM_006253569.2 | XP_006253631.1 | 4 | 983 | |||
| GenBank mRNA | BC061966.1 | - | 3 | 1165 | 5621094 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pitx1| Gene Symbol: Pitx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053624.2 | NP_446076.1 | 3 | 1165 | ||
| XM_006253569.2 | XP_006253631.1 | 4 | 983 | |||
| GenBank mRNA | BC061966.1 | - | 3 | 1165 | 5621094 | |