siRNA Details
| Gene Symbol: Pitx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053624.2 | NP_446076.1 | 1, 2 | 450 | ||
| XM_006253569.2 | XP_006253631.1 | 2, 3 | 268 | |||
| GenBank mRNA | AB047298.1 | - | 1, 2 | 471 | ||
| BC061966.1 | - | 1, 2 | 450 | 5621094 | ||

Pitx1
Genomic Map