siRNA Details
| Gene Symbol: Hmgn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007020.2 | NP_001007021.1 | 2 | 175 | ||
| NM_001201353.1 | NP_001188282.1 | 2 | 175 | |||
| NM_001201354.1 | NP_001188283.1 | 2 | 175 | |||
| NM_001201355.1 | NP_001188284.1 | 2 | 175 | |||
| XM_006243445.2 | XP_006243507.1 | 2 | 680 | |||
| XM_006243446.2 | XP_006243508.1 | 2 | 680 | |||
| XM_008766200.1 | XP_008764422.1 | 2 | 680 | |||
| GenBank mRNA | BC082036.1 | - | 2 | 130 | 7130894 | |
| CB751211.1 | - | 2 | 58 | |||
| FM058030.1 | - | 2 | 175 | |||

Hmgn3
Genomic Map