siRNA Details
| Gene Symbol: Cml5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080884.1 | NP_543160.1 | 2 | 1036 | ||
| XM_006236720.2 | XP_006236782.1 | 2 | 915 | |||
| XM_006236721.2 | XP_006236783.1 | 2 | 2444 | |||
| XM_006236722.2 | XP_006236784.1 | 2 | 941 | |||
| XM_006236723.1 | XP_006236785.1 | 1 | 680 | |||
| XM_006236725.2 | XP_006236787.1 | 2 | 699 | |||
| XM_008763044.1 | XP_008761266.1 | 3 | 1003 | |||
| GenBank mRNA | AF187100.1 | - | 1 | 368 | ||
| BC158648.1 | - | 2 | 1036 | 8367103 | ||

Cml5
Genomic Map