siRNA Details
| Gene Symbol: Ctnnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271502.1 | NP_001258431.1 | 11, 12 | 2113 | ||
| XM_006232094.2 | XP_006232156.1 | 11, 12 | 3090 | |||
| GenBank mRNA | AB008752.1 | - | 3, 4 | 478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnnd2| Gene Symbol: Ctnnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271502.1 | NP_001258431.1 | 11, 12 | 2113 | ||
| XM_006232094.2 | XP_006232156.1 | 11, 12 | 3090 | |||
| GenBank mRNA | AB008752.1 | - | 3, 4 | 478 | ||