siRNA Details
Gene Symbol: Fstl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053629.3 | NP_446081.1 | 2 | 182 | ||
GenBank mRNA | AB021295.1 | - | 2 | 182 | ||
AB071603.1 | - | 203 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fstl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053629.3 | NP_446081.1 | 2 | 182 | ||
GenBank mRNA | AB021295.1 | - | 2 | 182 | ||
AB071603.1 | - | 203 |