siRNA Details
| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053639.2 | NP_446091.2 | 2 | 117 | ||
| XM_006246219.2 | XP_006246281.1 | 2 | 310 | |||
| XM_008767655.1 | XP_008765877.1 | 3 | 1138 | |||
| GenBank mRNA | AB048790.2 | - | 2 | 117 | ||

Ltc4s
Genomic Map