siRNA Details
Gene Symbol: Ltc4s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053639.2 | NP_446091.2 | 3, 4 | 248 | ||
XM_006246219.2 | XP_006246281.1 | 3, 4 | 441 | |||
XM_008767655.1 | XP_008765877.1 | 4, 5 | 1269 | |||
GenBank mRNA | AB048790.2 | - | 3, 4 | 248 |
