siRNA Details
| Gene Symbol: Ltb4r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053640.1 | NP_446092.1 | 1 | 684 | ||
| XM_008770658.1 | XP_008768880.1 | 1 | 704 | |||
| GenBank mRNA | AB052660.1 | - | 1 | 684 | ||
More Information
Set Membership: |
|

Ltb4r2
Genomic Map