siRNA Details
| Gene Symbol: Bco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053648.3 | NP_446100.2 | 10, 11 | 1523 | ||
| GenBank mRNA | FQ219222.1 | - | 10, 11 | 1574 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bco1| Gene Symbol: Bco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053648.3 | NP_446100.2 | 10, 11 | 1523 | ||
| GenBank mRNA | FQ219222.1 | - | 10, 11 | 1574 | ||