siRNA Details
| Gene Symbol: Pafah1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053654.2 | NP_446106.1 | 4 | 309 | ||
| XM_006228365.2 | XP_006228427.1 | 4 | 447 | |||
| GenBank mRNA | AF016047.1 | - | 4 | 309 | ||
| BC126079.1 | - | 3 | 875 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pafah1b3| Gene Symbol: Pafah1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053654.2 | NP_446106.1 | 4 | 309 | ||
| XM_006228365.2 | XP_006228427.1 | 4 | 447 | |||
| GenBank mRNA | AF016047.1 | - | 4 | 309 | ||
| BC126079.1 | - | 3 | 875 | |||