siRNA Details
| Gene Symbol: Gng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053658.1 | NP_446110.1 | 2 | 284 | ||
| XM_008760212.1 | XP_008758434.1 | 1 | 366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gng3| Gene Symbol: Gng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053658.1 | NP_446110.1 | 2 | 284 | ||
| XM_008760212.1 | XP_008758434.1 | 1 | 366 | |||