siRNA Details
| Gene Symbol: Skil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232237.2 | XP_006232299.1 | 7 | 3031 | ||
| XM_008760948.1 | XP_008759170.1 | 6 | 4037 | |||
| XM_008760949.1 | XP_008759171.1 | 6 | 3897 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skil| Gene Symbol: Skil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232237.2 | XP_006232299.1 | 7 | 3031 | ||
| XM_008760948.1 | XP_008759170.1 | 6 | 4037 | |||
| XM_008760949.1 | XP_008759171.1 | 6 | 3897 | |||