siRNA Details
| Gene Symbol: Rbm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053696.1 | NP_446148.1 | 2, 3 | 199 | ||
| XM_006256694.1 | XP_006256756.1 | 2, 3 | 221 | |||
| XM_006256695.1 | XP_006256757.1 | 2, 3 | 221 | |||
| XM_006256696.1 | XP_006256758.1 | 2, 3 | 221 | |||
| GenBank mRNA | AF355190.1 | - | 2, 3 | 163 | ||
| BC087677.1 | - | 2, 3 | 156 | 7318884 | ||
| DN933310.1 | - | 1, 2 | 64 | |||
| DY319481.1 | - | 2, 3 | 199 | |||
| FQ210873.1 | - | 2, 3 | 178 | |||
| FQ220834.1 | - | 2, 3 | 174 | |||
| FQ231739.1 | - | 2, 3 | 174 | |||
| FQ232482.1 | - | 2, 3 | 177 | |||
| FQ235187.1 | - | 176 | ||||

Rbm3
Genomic Map