siRNA Details
| Gene Symbol: Ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053702.3 | NP_446154.3 | 4 | 704 | ||
| XM_006232266.2 | XP_006232328.1 | 5 | 841 | |||
| XM_008760903.1 | XP_008759125.1 | 5 | 836 | |||
| GenBank mRNA | AF367448.1 | - | 4 | 603 | ||
| AY623027.1 | - | 3 | 678 | |||
| AY623028.1 | - | 4 | 1083 | |||
| BC127462.1 | - | 5 | 797 | 8373429 | ||
| DY551463.1 | - | 4 | 415 | |||

Ccna2
Genomic Map