siRNA Details
| Gene Symbol: Klf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053713.1 | NP_446165.1 | 3 | 364 | ||
| GenBank mRNA | AF390546.1 | - | 3 | 364 | ||
| BC085720.1 | - | 3 | 757 | 7190219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf4| Gene Symbol: Klf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053713.1 | NP_446165.1 | 3 | 364 | ||
| GenBank mRNA | AF390546.1 | - | 3 | 364 | ||
| BC085720.1 | - | 3 | 757 | 7190219 | ||