siRNA Details
| Gene Symbol: Slc7a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053726.2 | NP_446178.2 | 7 | 999 | ||
| XM_006228873.2 | XP_006228935.1 | 7 | 1077 | |||
| XM_006228874.2 | XP_006228936.1 | 7 | 1077 | |||
| GenBank mRNA | AB126813.1 | - | 7 | 999 | ||
| BC127467.1 | - | 7 | 970 | 7313174 | ||

Slc7a10
Genomic Map