siRNA Details
| Gene Symbol: Ncf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053734.2 | NP_446186.2 | 4 | 389 | ||
| GenBank mRNA | AF260779.1 | - | 3 | 317 | ||
| AF547392.1 | - | 4 | 389 | |||
| AF547393.1 | - | 4 | 389 | |||
| AY029167.1 | - | 4 | 389 | |||
| BC061810.1 | - | 4 | 389 | 5598653 | ||
| DQ294708.1 | - | 1 | 146 | |||
| DQ294709.1 | - | 1 | 146 | |||

Ncf1
Genomic Map