siRNA Details
| Gene Symbol: Atp5g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053756.1 | NP_446208.1 | 4 | 540 | ||
| GenBank mRNA | AF315374.1 | - | 4 | 540 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5g3| Gene Symbol: Atp5g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053756.1 | NP_446208.1 | 4 | 540 | ||
| GenBank mRNA | AF315374.1 | - | 4 | 540 | ||