siRNA Details
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053760.2 | NP_446212.2 | 5 | 462 | ||
XM_006249376.2 | XP_006249438.1 | 5 | 518 | |||
XM_008769209.1 | XP_008767431.1 | 5 | 565 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053760.2 | NP_446212.2 | 5 | 462 | ||
XM_006249376.2 | XP_006249438.1 | 5 | 518 | |||
XM_008769209.1 | XP_008767431.1 | 5 | 565 |