siRNA Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053761.1 | NP_446213.1 | 5 | 1059 | ||
| XM_006236387.2 | XP_006236449.1 | 5 | 1214 | |||
| XM_006236388.2 | XP_006236450.1 | 5 | 1177 | |||
| XM_006236389.2 | XP_006236451.1 | 4 | 1121 | |||

Zyx
Genomic Map