siRNA Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053761.1 | NP_446213.1 | 7 | 1462 | ||
| XM_006236387.2 | XP_006236449.1 | 7 | 1617 | |||
| XM_006236388.2 | XP_006236450.1 | 7 | 1580 | |||
| XM_006236389.2 | XP_006236451.1 | 6 | 1524 | |||
| GenBank mRNA | FQ234073.1 | - | 1 | 59 | ||

Zyx
Genomic Map