siRNA Details
| Gene Symbol: Rgs14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053764.1 | NP_446216.1 | 2, 3 | 313 | ||
| GenBank mRNA | BC091132.1 | - | 1, 2 | 237 | 7376916 | |
| U92279.1 | - | 2, 3 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs14| Gene Symbol: Rgs14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053764.1 | NP_446216.1 | 2, 3 | 313 | ||
| GenBank mRNA | BC091132.1 | - | 1, 2 | 237 | 7376916 | |
| U92279.1 | - | 2, 3 | 313 | |||