siRNA Details
| Gene Symbol: Ifitm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030833.1 | NP_110460.1 | 1 | 102 | ||
| GenBank mRNA | AF164040.1 | - | 1 | 102 | ||
| BC060563.1 | - | 69 | 6918334 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm2| Gene Symbol: Ifitm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030833.1 | NP_110460.1 | 1 | 102 | ||
| GenBank mRNA | AF164040.1 | - | 1 | 102 | ||
| BC060563.1 | - | 69 | 6918334 | |||