siRNA Details
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105743.1 | NP_001099213.1 | 7 | 892 | ||
GenBank mRNA | AB001453.1 | - | 7 | 1106 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105743.1 | NP_001099213.1 | 7 | 892 | ||
GenBank mRNA | AB001453.1 | - | 7 | 1106 |