siRNA Details
| Gene Symbol: Gale | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080783.2 | NP_542961.2 | 10 | 942 | ||
| XM_006239134.2 | XP_006239196.1 | 10 | 1907 | |||
| XM_008764197.1 | XP_008762419.1 | 10 | 1173 | |||
| GenBank mRNA | BC097293.1 | - | 10 | 942 | 7443707 | |
| X53949.1 | - | 9 | 920 | |||

Gale
Genomic Map