siRNA Details
| Gene Symbol: Lim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053771.1 | NP_446223.1 | 2, 3 | 221 | ||
| GenBank mRNA | L04191.1 | - | 2, 3 | 199 | ||
| M87053.1 | - | 2, 3 | 221 | |||
| S55224.1 | - | 2, 3 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lim2| Gene Symbol: Lim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053771.1 | NP_446223.1 | 2, 3 | 221 | ||
| GenBank mRNA | L04191.1 | - | 2, 3 | 199 | ||
| M87053.1 | - | 2, 3 | 221 | |||
| S55224.1 | - | 2, 3 | 212 | |||