siRNA Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053773.1 | NP_446225.1 | 22 | 3299 | ||
| XM_006231175.2 | XP_006231237.1 | 22 | 4381 | |||
| XM_006231176.2 | XP_006231238.1 | 21 | 4271 | |||
| XM_006231177.2 | XP_006231239.1 | 21 | 3612 | |||
| XM_006231178.2 | XP_006231240.1 | 21 | 6541 | |||
| XM_006231179.2 | XP_006231241.1 | 21 | 3445 | |||
| XM_006231181.2 | XP_006231243.1 | 22 | 3400 | |||
| XM_006231182.2 | XP_006231244.1 | 20 | 3501 | |||
| XM_008760302.1 | XP_008758524.1 | 22 | 3141 | |||
| GenBank mRNA | BC103479.1 | - | 22 | 3299 | 7936785 | |

Tjp2
Genomic Map