siRNA Details
| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053775.3 | NP_446227.2 | 4 | 329 | ||
| GenBank mRNA | AF118854.1 | - | 1 | 47 | ||
| BC168659.1 | - | 414 | 7938299 | |||
| DQ286425.1 | - | 4 | 329 | |||
| DV216525.1 | - | 4 | 417 | |||

Atp6v0a2
Genomic Map