siRNA Details
| Gene Symbol: Ptcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750652.3 | XP_003750700.1 | 2 | 921 | ||
| XM_008766898.1 | XP_008765120.1 | 2 | 568 | |||
| GenBank mRNA | AF182508.1 | - | 1 | 140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptcra| Gene Symbol: Ptcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750652.3 | XP_003750700.1 | 2 | 921 | ||
| XM_008766898.1 | XP_008765120.1 | 2 | 568 | |||
| GenBank mRNA | AF182508.1 | - | 1 | 140 | ||