siRNA Details
| Gene Symbol: Ptcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750652.3 | XP_003750700.1 | 4 | 1151 | ||
| XM_008766898.1 | XP_008765120.1 | 4 | 798 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptcra| Gene Symbol: Ptcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750652.3 | XP_003750700.1 | 4 | 1151 | ||
| XM_008766898.1 | XP_008765120.1 | 4 | 798 | |||