siRNA Details
| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053787.2 | NP_446239.2 | 8 | 1242 | ||
| XM_006236376.2 | XP_006236438.1 | 6 | 818 | |||
| GenBank mRNA | AB032019.1 | - | 8 | 1242 | ||
| AF209196.1 | - | 1193 | ||||
| AY762624.1 | - | 7 | 930 | |||

Trpv5
Genomic Map