siRNA Details
| Gene Symbol: Il17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053789.1 | NP_446241.1 | 3 | 568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17b| Gene Symbol: Il17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053789.1 | NP_446241.1 | 3 | 568 | ||