siRNA Details
| Gene Symbol: Kcnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053804.2 | NP_446256.2 | 6 | 910 | ||
| XM_006230634.2 | XP_006230696.1 | 7 | 2208 | |||
| XM_008760054.1 | XP_008758276.1 | 7 | 1951 | |||
| XM_008760055.1 | XP_008758277.1 | 7 | 1822 | |||
| XM_008760056.1 | XP_008758278.1 | 7 | 1028 | |||
| GenBank mRNA | AF259502.1 | - | 2 | 276 | ||
| AF302842.2 | - | 6 | 910 | |||

Kcnk4
Genomic Map