siRNA Details
| Gene Symbol: Snai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053805.1 | NP_446257.1 | 2 | 541 | ||
| GenBank mRNA | BC079210.1 | - | 2 | 541 | 7131806 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snai1| Gene Symbol: Snai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053805.1 | NP_446257.1 | 2 | 541 | ||
| GenBank mRNA | BC079210.1 | - | 2 | 541 | 7131806 | |