siRNA Details
| Gene Symbol: Fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053809.1 | NP_446261.1 | 3 | 551 | ||
| XM_006230640.2 | XP_006230702.1 | 3 | 681 | |||
| GenBank mRNA | AB079673.1 | - | 3 | 551 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf4| Gene Symbol: Fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053809.1 | NP_446261.1 | 3 | 551 | ||
| XM_006230640.2 | XP_006230702.1 | 3 | 681 | |||
| GenBank mRNA | AB079673.1 | - | 3 | 551 | ||