siRNA Details
| Gene Symbol: Ebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053820.2 | NP_446272.2 | 10, 11 | 1297 | ||
| XM_006246117.2 | XP_006246179.1 | 10, 11 | 2902 | |||
| XM_006246118.2 | XP_006246180.1 | 10, 11 | 2899 | |||
| XM_006246120.2 | XP_006246182.1 | 11, 12 | 2656 | |||
| XM_008767631.1 | XP_008765853.1 | 10, 11 | 1253 | |||
| XM_008767632.1 | XP_008765854.1 | 10, 11 | 1667 | |||
| XM_008767633.1 | XP_008765855.1 | 8, 9 | 788 | |||
| XM_008767634.1 | XP_008765856.1 | 7, 8 | 1150 | |||
| GenBank mRNA | L24051.1 | - | 10, 11 | 1039 | ||

Ebf1
Genomic Map