siRNA Details
| Gene Symbol: Pdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053826.2 | NP_446278.2 | 4 | 576 | ||
| XM_006234352.2 | XP_006234414.1 | 4 | 348 | |||
| GenBank mRNA | BC089783.1 | - | 4 | 576 | 7321326 | |
| L22294.1 | - | 4 | 507 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdk1| Gene Symbol: Pdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053826.2 | NP_446278.2 | 4 | 576 | ||
| XM_006234352.2 | XP_006234414.1 | 4 | 348 | |||
| GenBank mRNA | BC089783.1 | - | 4 | 576 | 7321326 | |
| L22294.1 | - | 4 | 507 | |||