siRNA Details
| Gene Symbol: Il13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053828.1 | NP_446280.1 | 1 | 111 | ||
| GenBank mRNA | L26913.1 | - | 1 | 111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il13| Gene Symbol: Il13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053828.1 | NP_446280.1 | 1 | 111 | ||
| GenBank mRNA | L26913.1 | - | 1 | 111 | ||