siRNA Details
| Gene Symbol: Kcnj9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053834.1 | NP_446286.1 | 3 | 989 | ||
| GenBank mRNA | L77929.1 | - | 3 | 989 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj9| Gene Symbol: Kcnj9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053834.1 | NP_446286.1 | 3 | 989 | ||
| GenBank mRNA | L77929.1 | - | 3 | 989 | ||