siRNA Details
| Gene Symbol: Nat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037315.1 | NP_001032392.1 | 3 | 591 | ||
| NM_001037316.1 | NP_001032393.1 | 2 | 349 | |||
| NM_053853.2 | NP_446305.1 | 1 | 744 | |||
| XM_006253001.1 | XP_006253063.1 | 3 | 415 | |||
| XM_006253002.2 | XP_006253064.1 | 1 | 274 | |||
| XM_006253003.1 | XP_006253065.1 | 3 | 419 | |||
| XM_006253004.2 | XP_006253066.1 | 3 | 630 | |||
| GenBank mRNA | BC078765.1 | - | 2 | 350 | 7127165 | |
| U01343.1 | - | 3 | 591 | |||
| U01344.1 | - | 1 | 1242 | |||
| U17260.1 | - | 2 | 411 | |||

Nat1
Genomic Map