siRNA Details
| Gene Symbol: E2f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760849.1 | XP_008759071.1 | 5 | 1719 | ||
| XM_574892.6 | XP_574892.1 | 6 | 1190 | |||
| GenBank mRNA | U31668.1 | - | 6 | 539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E2f5| Gene Symbol: E2f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760849.1 | XP_008759071.1 | 5 | 1719 | ||
| XM_574892.6 | XP_574892.1 | 6 | 1190 | |||
| GenBank mRNA | U31668.1 | - | 6 | 539 | ||