siRNA Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053877.3 | NP_446329.1 | 6, 7 | 483 | ||
| GenBank mRNA | BC061527.1 | - | 6, 7 | 484 | 6920755 | |
| U34843.1 | - | 6, 7 | 495 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc123| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053877.3 | NP_446329.1 | 6, 7 | 483 | ||
| GenBank mRNA | BC061527.1 | - | 6, 7 | 484 | 6920755 | |
| U34843.1 | - | 6, 7 | 495 | |||