siRNA Details
| Gene Symbol: Vwf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053889.1 | NP_446341.1 | 35, 36 | 6290 | ||
| XM_008763279.1 | XP_008761501.1 | 35, 36 | 6279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwf| Gene Symbol: Vwf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053889.1 | NP_446341.1 | 35, 36 | 6290 | ||
| XM_008763279.1 | XP_008761501.1 | 35, 36 | 6279 | |||