siRNA Details
| Gene Symbol: Cdk5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053891.1 | NP_446343.1 | 1 | 368 | ||
| GenBank mRNA | U50707.1 | - | 1 | 368 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk5r1| Gene Symbol: Cdk5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053891.1 | NP_446343.1 | 1 | 368 | ||
| GenBank mRNA | U50707.1 | - | 1 | 368 | ||