siRNA Details
| Gene Symbol: Kcnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105748.1 | NP_001099218.1 | 5, 6 | 1724 | ||
| GenBank mRNA | U89873.1 | - | 5, 6 | 1269 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnd1| Gene Symbol: Kcnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105748.1 | NP_001099218.1 | 5, 6 | 1724 | ||
| GenBank mRNA | U89873.1 | - | 5, 6 | 1269 | ||