siRNA Details
| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053931.3 | NP_446383.3 | 2 | 357 | ||
| GenBank mRNA | AB027143.1 | - | 2 | 357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sept5| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053931.3 | NP_446383.3 | 2 | 357 | ||
| GenBank mRNA | AB027143.1 | - | 2 | 357 | ||